Learn More
Medchemexpress LLC Methyl methanesulfonate (Standard) | 66-27-3 | 110.13 | 250 MG
SDP
Catalog No. 5000450061
Encompass_Preferred
Methyl methanesulfonate (Standard) is the analytical standard of Methyl methanesulfonate. This product is intended for research and analytical applications. It is an alkylating agent which transfers methyl groups and induces DNA damage. It is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • The compound is an analytical standard, which is a reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-50061 Supplier Medchemexpress LLC Supplier No. 250MGHYW004702R
Methyl methanesulfonate (Standard) is the analytical standard of Methyl methanesulfonate. This product is intended for research and analytical applications. It is an alkylating agent which transfers methyl groups and induces DNA damage. It is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • The compound is an analytical standard, which is a reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.