Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl methylphenylphosphinate, 98%

Catalog No. AAA1876006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1876006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1876006 Supplier Thermo Scientific Chemicals Supplier No. A1876006
Add to Cart

CAS: 6389-79-3 | C8H11O2P | 170.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6389-79-3
Molecular Formula C8H11O2P
Molecular Weight (g/mol) 170.15
MDL Number MFCD00013909
InChI Key RTAAKNLNWIHGGX-UHFFFAOYNA-N
Synonym methyl methylphenylphosphinate, methyl methyl phenyl phosphinate, methoxy methyl phosphoryl benzene, acmc-1b8qc, methylphenylphosphinic acid methyl ester, phenyl methyl phosphinic acid methyl ester, +-methylphenylphosphinic acid methyl ester, ?-methylphenylphosphinic acid methyl ester, phosphinic acid, methylphenyl-, methyl ester, r
PubChem CID 323835
IUPAC Name [methoxy(methyl)phosphoryl]benzene
SMILES COP(C)(=O)C1=CC=CC=C1

Specifications

Boiling Point 148°C to 149°C (17 mmHg)
Quantity 5 g
Formula Weight 170.15
Percent Purity 98%
Chemical Name or Material Methyl methylphenylphosphinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.