Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl methylsulfonylacetate, 98+%

Catalog No. AAL0803906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0803906 5 g
AAL0803914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0803906 Supplier Thermo Scientific Chemicals Supplier No. L0803906
Add to Cart

CAS: 62020-09-1 | C4H8O4S | 152.16 g/mol

Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62020-09-1
Molecular Formula C4H8O4S
Molecular Weight (g/mol) 152.16
MDL Number MFCD00051842
InChI Key RQKDASVRZONLNQ-UHFFFAOYSA-N
Synonym methyl methanesulfonylacetate, methyl 2-methanesulfonylacetate, methyl 2-methylsulfonyl acetate, methyl methylsulfonylacetate, methanesulfonylacetic acid methyl ester, acmc-1b4fd, methyl methylsulfonyl acetate, 2-methylsulfonyl-1-methoxyethanone, methylsulfonylacetic acid methyl ester, methanesulfonyl acetic acid methyl ester
PubChem CID 2759918
SMILES COC(=O)CS(C)(=O)=O

Specifications

Melting Point 59°C to 63°C
Quantity 5 g
Beilstein 2412782
Solubility Information Soluble in methanol. Insoluble in water.
IUPAC Name methyl 2-methanesulfonylacetate
Formula Weight 152.17
Percent Purity ≥98%
Chemical Name or Material Methyl methylsulfonylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.