Learn More
Medchemexpress LLC Methyl mycophenolate | 31858-66-9 | 99.0% | 334.37 | 1 ML
SDP
Catalog No. 5000437265
Encompass_Preferred
Methyl mycophenolate is a methyl ester of mycophenolic acid also found in the marine-derived fungus *Phaeosphaeria spartinae*. It is intended for research use only.
  • Cytotoxicity against various human cell lines.
  • Soluble in DMSO.
  • Available in solid and solution formulations.
Catalog No. 50-004-37265 Supplier Medchemexpress LLC Supplier No. HY11397210MM/1ML
Methyl mycophenolate is a methyl ester of mycophenolic acid also found in the marine-derived fungus *Phaeosphaeria spartinae*. It is intended for research use only.
  • Cytotoxicity against various human cell lines.
  • Soluble in DMSO.
  • Available in solid and solution formulations.