Learn More
Medchemexpress LLC Methyl mycophenolate | 31858-66-9 | 99.0% | 334.37 | 25 MG
SDP
Catalog No. 5000437617
Encompass_Preferred
Methyl mycophenolate is a methyl ester of mycophenolic acid. It is found in the marine-derived fungus Phaeosphaeria spartinae and is for research use only.
  • Methyl ester of mycophenolic acid
  • Found in marine-derived fungus Phaeosphaeria spartinae
  • For research use only
Catalog No. 50-004-37617 Supplier Medchemexpress LLC Supplier No. 25MGHY113972
Methyl mycophenolate is a methyl ester of mycophenolic acid. It is found in the marine-derived fungus Phaeosphaeria spartinae and is for research use only.
  • Methyl ester of mycophenolic acid
  • Found in marine-derived fungus Phaeosphaeria spartinae
  • For research use only