Learn More
Medchemexpress LLC HY-113972 5mg Medchemexpress, Methyl mycophenolate CAS:31858-66-9 Purity:>98%
SDP
Catalog No. 501969277
Encompass_Preferred
Medchemexpress, HY-113972 5mg Methyl mycophenolate CAS:31858-66-9 Methyl mycophenolate is a methyl ester of mycophenolic acid and is also found in marine-derived fungus Phaeosphaeria spartinae. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9277 Supplier Medchemexpress LLC Supplier No. HY1139725MG
Medchemexpress, HY-113972 5mg Methyl mycophenolate CAS:31858-66-9 Methyl mycophenolate is a methyl ester of mycophenolic acid and is also found in marine-derived fungus Phaeosphaeria spartinae. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.