Learn More
Medchemexpress LLC Methyl myristate | 124-10-7 | 242.40 | 1 G
SDP
Catalog No. 5000446933
Encompass_Preferred
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It induces high melanin in B16F10 melanoma. This compound is also a component of petroleum ether extract, which reduces Acetylcholine and ergot alkaloids-induced uterine contractions in rats isolated uterus, exhibiting uterine relaxation activity.
  • Saturated fatty acid methyl ester
  • Obtained from the esterification of myristic acid
  • Induces high melanin in B16F10 melanoma
  • Component of petroleum ether extract
  • Exhibits uterine relaxation activity in isolated rat uterus
Catalog No. 50-004-46933 Supplier Medchemexpress LLC Supplier No. HYW0042881G
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It induces high melanin in B16F10 melanoma. This compound is also a component of petroleum ether extract, which reduces Acetylcholine and ergot alkaloids-induced uterine contractions in rats isolated uterus, exhibiting uterine relaxation activity.
  • Saturated fatty acid methyl ester
  • Obtained from the esterification of myristic acid
  • Induces high melanin in B16F10 melanoma
  • Component of petroleum ether extract
  • Exhibits uterine relaxation activity in isolated rat uterus