Learn More
Medchemexpress LLC Methyl myristate | 124-10-7 | 242.40 | 10 G
SDP
Catalog No. 5000445457
Encompass_Preferred
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It demonstrates a high melanin induction in B16F10 melanoma. It is also a component of petroleum ether extract, which has shown uterine relaxation activity by reducing acetylcholine and ergot alkaloids-induced uterine contractions in isolated rat uteri.
  • Derived from myristic acid
  • Shows high melanin induction in B16F10 melanoma
  • Component of petroleum ether extract
  • Exhibits uterine relaxation activity
Catalog No. 50-004-45457 Supplier Medchemexpress LLC Supplier No. HYW00428810G
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It demonstrates a high melanin induction in B16F10 melanoma. It is also a component of petroleum ether extract, which has shown uterine relaxation activity by reducing acetylcholine and ergot alkaloids-induced uterine contractions in isolated rat uteri.
  • Derived from myristic acid
  • Shows high melanin induction in B16F10 melanoma
  • Component of petroleum ether extract
  • Exhibits uterine relaxation activity