Learn More
Medchemexpress LLC Methyl myristate | 124-10-7 | 242.40 | 5 G
SDP
Catalog No. 5000448802
Encompass_Preferred
Methyl myristate is a saturated fatty acid methyl ester produced through the esterification of myristic acid. It has been observed to exhibit a high melanin induction in B16F10 melanoma cells.
  • In vitro activity: Methyl myristate is a component of petroleum ether extract.
  • This extract has demonstrated the ability to reduce acetylcholine and ergot alkaloid-induced uterine contractions in isolated rat uteri, indicating uterine relaxation activity.
Catalog No. 50-004-48802 Supplier Medchemexpress LLC Supplier No. HYW0042885G
Methyl myristate is a saturated fatty acid methyl ester produced through the esterification of myristic acid. It has been observed to exhibit a high melanin induction in B16F10 melanoma cells.
  • In vitro activity: Methyl myristate is a component of petroleum ether extract.
  • This extract has demonstrated the ability to reduce acetylcholine and ergot alkaloid-induced uterine contractions in isolated rat uteri, indicating uterine relaxation activity.