Learn More
Medchemexpress LLC Methyl myristate | 124-10-7 | MFCD00008983 | 242.40 | 1 ML
SDP
Catalog No. 5000445455
Encompass_Preferred
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It demonstrates significant melanin induction in B16F10 melanoma.
  • High melanin induction in B16F10 melanoma.
  • Available in a liquid and solvent form (10 mM in 1 mL of DMSO), ready for reconstitution.
Catalog No. 50-004-45455 Supplier Medchemexpress LLC Supplier No. HYW00428810MM/1ML
Methyl myristate is a saturated fatty acid methyl ester obtained from the esterification of myristic acid. It demonstrates significant melanin induction in B16F10 melanoma.
  • High melanin induction in B16F10 melanoma.
  • Available in a liquid and solvent form (10 mM in 1 mL of DMSO), ready for reconstitution.