Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl myristelaidate, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015343 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5343 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5343 Supplier MilliporeSigma Supelco Supplier No. 70055100MG
Add to Cart
Add to Cart

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 72025-18-4
Shipping Condition Ambient
Physical Form Neat
Linear Formula C15H28O2
MDL Number MFCD00133548
Synonym Methyl trans-9-tetradecenoate; Myristelaidic acid methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 240.38
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.