Learn More
Medchemexpress LLC Methyl N-[[(5S)-3-[3-fluoro-4-(2-oxa-6-azaspiro[3.3]heptan-6-yl)phenyl]-2-oxo-1,3-oxazolidin-5-yl... | 2071265-08-0 | 99.7% | C17H20FN3O5 | 10MG
SDP
Catalog No. 5000261284
Encompass_Preferred
TBI-223 is an orally active oxazolidinone-class antibiotic research compound showing activity against Mycobacterium tuberculosis and demonstrated efficacy in mouse infection models. It inhibits mitochondrial protein synthesis in HepG2 cells and is supplied as a solid with defined storage recommendations for long-term stability.
  • Orally active oxazolidinone scaffold.
  • Shows activity against Mycobacterium tuberculosis.
  • Demonstrated efficacy in multiple mouse infection models.
  • Inhibits mitochondrial protein synthesis in HepG2 cells (IC50 = 68 μg/mL).
  • High purity and defined storage conditions for stability.
Catalog No. 50-002-61284 Supplier Medchemexpress LLC Supplier No. HY13939810MG
TBI-223 is an orally active oxazolidinone-class antibiotic research compound showing activity against Mycobacterium tuberculosis and demonstrated efficacy in mouse infection models. It inhibits mitochondrial protein synthesis in HepG2 cells and is supplied as a solid with defined storage recommendations for long-term stability.
  • Orally active oxazolidinone scaffold.
  • Shows activity against Mycobacterium tuberculosis.
  • Demonstrated efficacy in multiple mouse infection models.
  • Inhibits mitochondrial protein synthesis in HepG2 cells (IC50 = 68 μg/mL).
  • High purity and defined storage conditions for stability.