Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl N-benzylpyrrolidine-3-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH3334103
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3334103 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3334103 Supplier Thermo Scientific Chemicals Supplier No. H3334103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 17012-21-4
Boiling Point 98°C (0.2mmHg)
Molecular Formula C13H17NO2
Refractive Index 1.5211
Quantity 1g
MDL Number MFCD04038678
Synonym methyl n-benzyl-3-pyrrolidinecarboxylate, 1-benzyl-pyrrolidine-3-carboxylic acid methyl ester, methyl 1-benzyl-3-pyrrolidinecarboxylate, methyl-1-benzylpyrrolidine-3carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, methyl ester, methyl 1-phenylmethyl pyrrolidine-3-carboxylate, methyl n-benzylpyrrolidine-3-carboxylate, methyl 1-benzyllpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid methyl ester, 1-phenylmethyl-3-pyrrolidinecarboxylic acid methyl ester
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
InChI Key GDWFCUOFVSNTTG-UHFFFAOYSA-N
SMILES COC(=O)C1CCN(C1)CC2=CC=CC=C2
IUPAC Name methyl 1-benzylpyrrolidine-3-carboxylate
Molecular Weight (g/mol) 219.284
PubChem CID 4610751
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material Methyl N-benzylpyrrolidine-3-carboxylate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.