Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl n-propyl sulfide, 99%, Thermo Scientific™

Catalog No. AAL0934618
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0934618 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0934618 Supplier Thermo Scientific Chemicals Supplier No. L0934618

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3877-15-4
Boiling Point 94°C to 96°C
Flash Point 11°C (51°F)
Odor Stench
Molecular Formula C4H10S
Quantity 50g
MDL Number MFCD00015219
UN Number UN1993
Beilstein 1730904
Synonym methyl propyl sulfide, propane, 1-methylthio, sulfide, methyl propyl, methyl propyl sulphide, unii-nxa8rq27b3, 1-methylthio propane, 1-methylthio-propane, n-propylmethylsulfide, nxa8rq27b3, methyl n-propyl sulfide
InChI Key ZOASGOXWEHUTKZ-UHFFFAOYSA-N
SMILES CCCSC
IUPAC Name 1-methylsulfanylpropane
Molecular Weight (g/mol) 90.18
PubChem CID 19754
Formula Weight 90.19
Percent Purity 99%
Chemical Name or Material Methyl n-propyl sulfide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.