Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate 98.0+%, TCI America™
SDP

Catalog No. M28035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28035G 5 g
M280325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28035G Supplier TCI America Supplier No. M28035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108963-96-8
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD06809720
InChI Key FNTAOUUEQHKLIU-ZETCQYMHSA-N
Synonym boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135
PubChem CID 10868485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC

Specifications

Melting Point 71°C
Color White
Quantity 5 g
Formula Weight 243.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.