Learn More
Medchemexpress LLC Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate | 134167-07-0 | MFCD18837831 | >95.0% | 233.26 g/mol | C10H19NO5 | 5 G
SDP
Catalog No. 5000209925
Encompass_Preferred
Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is an N-Boc protected serine methyl ester used as an intermediate and building block in organic synthesis and biochemical research. It is supplied for research use only.
  • N-Boc protected serine derivative useful as a building block for peptide and small-molecule synthesis.
  • O-methylated side chain enables selective functionalization of the serine hydroxyl position.
  • Available in multiple pack sizes for lab-scale synthesis and method development.
  • Stable under standard laboratory storage conditions; store as recommended on safety documentation.
Catalog No. 50-002-09925 Supplier Medchemexpress LLC Supplier No. HYW0574345G
Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is an N-Boc protected serine methyl ester used as an intermediate and building block in organic synthesis and biochemical research. It is supplied for research use only.
  • N-Boc protected serine derivative useful as a building block for peptide and small-molecule synthesis.
  • O-methylated side chain enables selective functionalization of the serine hydroxyl position.
  • Available in multiple pack sizes for lab-scale synthesis and method development.
  • Stable under standard laboratory storage conditions; store as recommended on safety documentation.