Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate | 134167-07-0 | MFCD18837831 | 97.0% | 233.26 g/mol | C10H19NO5 | 10 G
SDP

Catalog No. 5000210008
Encompass_Preferred

Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is a Boc-protected O-methyl derivative of L-serine used as an intermediate in peptide and organic synthesis. It is supplied as a solid with defined purity and storage recommendations.

  • Boc-protected O-methyl L-serine methyl ester.
  • Used as an intermediate in peptide and organic synthesis.
  • Molecular formula: C10H19NO5.
  • Molecular weight: 233.26 g/mol.
  • Purity: 97.0%.
  • Storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g (quote).

Catalog No. 50-002-10008 Supplier Medchemexpress LLC Supplier No. HYW05743410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is a Boc-protected O-methyl derivative of L-serine used as an intermediate in peptide and organic synthesis. It is supplied as a solid with defined purity and storage recommendations.

  • Boc-protected O-methyl L-serine methyl ester.
  • Used as an intermediate in peptide and organic synthesis.
  • Molecular formula: C10H19NO5.
  • Molecular weight: 233.26 g/mol.
  • Purity: 97.0%.
  • Storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g (quote).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.