Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate | 134167-07-0 | MFCD18837831 | 97.0% | 233.26 g/mol | C10H19NO5 | 250 MG
SDP

Catalog No. 5000209659
Encompass_Preferred

Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is a Boc-protected, O-methylated methyl ester of L-serine used as a research reagent and synthetic intermediate in organic and peptide synthesis.

  • Purity 97.0%.
  • Molecular weight 233.26 g/mol.
  • Chemical formula C10H19NO5.
  • Cas number 134167-07-0.
  • Available sizes 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g.
  • Storage pure form: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-09659 Supplier Medchemexpress LLC Supplier No. 250MGHYW057434
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl N-(tert-butoxycarbonyl)-O-methyl-L-serinate is a Boc-protected, O-methylated methyl ester of L-serine used as a research reagent and synthetic intermediate in organic and peptide synthesis.

  • Purity 97.0%.
  • Molecular weight 233.26 g/mol.
  • Chemical formula C10H19NO5.
  • Cas number 134167-07-0.
  • Available sizes 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g.
  • Storage pure form: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.