Learn More
Medchemexpress LLC Methyl N-(triethylammoniosulfonyl)carbamate (inner salt) | 29684-56-8 | MFCD00077815 | >95.0% | 238.31 g·mol^-1 | C8H18N2O4S | 25 G
SDP
Catalog No. 5000303526
Encompass_Preferred
Burgess reagent is a mild, selective dehydrating reagent used in organic synthesis to convert secondary and tertiary alcohols to alkenes under relatively mild conditions. It is typically handled as a crystalline powder (white to cream to yellow) and is chosen when selectivity and functional-group tolerance are important. Purity and appearance can vary by supplier and grade.
Catalog No. 50-003-03526 Supplier Medchemexpress LLC Supplier No. 25GHYW040081
Burgess reagent is a mild, selective dehydrating reagent used in organic synthesis to convert secondary and tertiary alcohols to alkenes under relatively mild conditions. It is typically handled as a crystalline powder (white to cream to yellow) and is chosen when selectivity and functional-group tolerance are important. Purity and appearance can vary by supplier and grade.