Learn More
Medchemexpress LLC HY-B1695 100mg Medchemexpress, Methyl nicotinate CAS:93-60-7 Purity:>98%
SDP
Catalog No. 501932990
Encompass_Preferred
Medchemexpress, HY-B1695 100mg Methyl nicotinate CAS:93-60-7 Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, that is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2990 Supplier Medchemexpress LLC Supplier No. HYB1695100MG
Medchemexpress, HY-B1695 100mg Methyl nicotinate CAS:93-60-7 Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, that is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.