Learn More
Selleck Chemical LLC Methyl nicotinate
SDP
Catalog No. 5000002965
Encompass_Preferred
Methyl nicotinate (methyl pyridine-3-carboxylate) is an ester of methyl alcohol and nicotinic acid It is a rubefacient
Catalog No. 50-000-02965 Supplier Selleck Chemical LLC Supplier No. S623125MG
Methyl nicotinate (methyl pyridine-3-carboxylate) is an ester of methyl alcohol and nicotinic acid It is a rubefacient