Learn More
Medchemexpress LLC Methyl nicotinate | 93-60-7 | 99.9% | 1 ML
SDP
Catalog No. 5000413452
Encompass_Preferred
Methyl nicotinate (Nicotinic acid methyl ester) is an orally active vasodilator with analgesic activity. It causes local skin erythema when applied topically and is used as an active ingredient in over-the-counter topical preparations for studying muscle and joint pain.
  • Orally active vasodilator.
  • Possesses analgesic activity.
  • Causes local skin erythema when applied topically.
  • Used in over-the-counter topical preparations for muscle and joint pain research.
Catalog No. 50-004-13452 Supplier Medchemexpress LLC Supplier No. HYB169510MM/1ML
Add to Cart
Add to Cart
Methyl nicotinate (Nicotinic acid methyl ester) is an orally active vasodilator with analgesic activity. It causes local skin erythema when applied topically and is used as an active ingredient in over-the-counter topical preparations for studying muscle and joint pain.
  • Orally active vasodilator.
  • Possesses analgesic activity.
  • Causes local skin erythema when applied topically.
  • Used in over-the-counter topical preparations for muscle and joint pain research.