Learn More
Medchemexpress LLC Methyl nicotinate | 93-60-7 | 99.95% | 25 G
SDP
Catalog No. 5000414191
Encompass_Preferred
Methyl nicotinate (Nicotinic acid methyl ester) is an orally active vasodilator with analgesic activity. It causes local skin erythema when applied topically and is used as an active ingredient in over-the-counter topical preparations for the study of muscle and joint pain.
  • Orally active vasodilator.
  • Exhibits analgesic activity.
  • Causes local skin erythema when applied topically.
  • Used as an active ingredient in over-the-counter topical preparations for muscle and joint pain research.
  • Shows peripheral and central antialgesic effects in mice.
  • Demonstrates a vasodilatory effect in egg embryo eggs.
Catalog No. 50-004-14191 Supplier Medchemexpress LLC Supplier No. 25GHYB1695
Methyl nicotinate (Nicotinic acid methyl ester) is an orally active vasodilator with analgesic activity. It causes local skin erythema when applied topically and is used as an active ingredient in over-the-counter topical preparations for the study of muscle and joint pain.
  • Orally active vasodilator.
  • Exhibits analgesic activity.
  • Causes local skin erythema when applied topically.
  • Used as an active ingredient in over-the-counter topical preparations for muscle and joint pain research.
  • Shows peripheral and central antialgesic effects in mice.
  • Demonstrates a vasodilatory effect in egg embryo eggs.