Learn More
Medchemexpress LLC Methyl nicotinate-d4 | 345909-99-1 | 99.5% | 100 MG
SDP
Catalog No. 5000412360
Encompass_Preferred
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate. Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12360 Supplier Medchemexpress LLC Supplier No. HYB1695S100MG
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate. Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.