Learn More
Medchemexpress LLC Methyl nicotinate-d4 | 345909-99-1 | 99.5% | 25 MG
SDP
Catalog No. 5000413845
Encompass_Preferred
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate. The non-deuterated Methyl nicotinate, the methyl ester of Niacin, is found in alcoholic beverages and is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Appearance: solid
  • Color: off-white to light yellow
Catalog No. 50-004-13845 Supplier Medchemexpress LLC Supplier No. 25MGHYB1695S
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate. The non-deuterated Methyl nicotinate, the methyl ester of Niacin, is found in alcoholic beverages and is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Appearance: solid
  • Color: off-white to light yellow