Learn More
Medchemexpress LLC Methyl nicotinate-d4 | 345909-99-1 | 99.5% | 5 MG
SDP
Catalog No. 5000413152
Encompass_Preferred
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate, used as an active ingredient as a rubefacient in over-the-counter topical preparations for muscle and joint pain. This compound can be used as a tracer and as an internal standard for quantitative analysis by various methods.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Methyl nicotinate is found in alcoholic beverages
  • Applications in muscle and joint pain preparations
Catalog No. 50-004-13152 Supplier Medchemexpress LLC Supplier No. HYB1695S5MG
Methyl nicotinate-d4 is the deuterium labeled Methyl nicotinate, used as an active ingredient as a rubefacient in over-the-counter topical preparations for muscle and joint pain. This compound can be used as a tracer and as an internal standard for quantitative analysis by various methods.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Methyl nicotinate is found in alcoholic beverages
  • Applications in muscle and joint pain preparations