Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl nicotinate-d4 | 345909-99-1 | MFCD01074082 | 99.5% | 141.16 g·mol-1 | C7H3D4NO2 | 10 MG
SDP

Catalog No. 5000298109
Encompass_Preferred

Methyl nicotinate-d4 is a deuterium-labeled analogue of methyl nicotinate supplied for research and analytical applications. It is provided as a high-purity solid intended for use as a tracer or internal standard in mass spectrometry and quantitative assays.

  • High isotopic enrichment and reported purity of 99.5%.
  • Molecular formula C7H3D4NO2; molecular weight ~141.16 g·mol-1.
  • Available in small mass pack sizes tailored for analytical workflows.
  • Solid appearance with recommended cold storage to maintain stability.
  • Intended for research use and analytical applications.

Catalog No. 50-002-98109 Supplier Medchemexpress LLC Supplier No. HYB1695S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl nicotinate-d4 is a deuterium-labeled analogue of methyl nicotinate supplied for research and analytical applications. It is provided as a high-purity solid intended for use as a tracer or internal standard in mass spectrometry and quantitative assays.

  • High isotopic enrichment and reported purity of 99.5%.
  • Molecular formula C7H3D4NO2; molecular weight ~141.16 g·mol-1.
  • Available in small mass pack sizes tailored for analytical workflows.
  • Solid appearance with recommended cold storage to maintain stability.
  • Intended for research use and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.