Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nicotinate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111001819 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1819 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1819 Supplier MilliporeSigma Supelco Supplier No. PHR1835500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methyl Nicotinate is a volatile product of certain fruits such as carambola, soursop, wild strawberries, etc. It is used for a variety of applications in food products, cosmetics, and medicinal products.
TRUSTED_SUSTAINABILITY

Specifications

CAS 93-60-7
Flash Point 96°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H7 NO2
MDL Number MFCD00006388
Melting Point 42°C to 44°C
Boiling Point 204°C
Synonym Methyl nicotinate; Nicotinic acid methyl ester
Type Certified Reference Material
Quantity 500 mg
Formula Weight 137.14
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.