Learn More
Selleck Chemical LLC Methyl Nonadecanoate
SDP
Catalog No. 5000003929
Encompass_Preferred
Methyl nonadecanoate (Nonadecanoic Acid methyl ester) is a fatty acid methyl ester which is less water soluble but more amenable for the formulation of nonadecanoate-containing diets and dietary supplements
Catalog No. 50-000-03929 Supplier Selleck Chemical LLC Supplier No. S583325MG
Methyl nonadecanoate (Nonadecanoic Acid methyl ester) is a fatty acid methyl ester which is less water soluble but more amenable for the formulation of nonadecanoate-containing diets and dietary supplements