Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nonafluorobutyl Ketone 99.0+%, TCI America™
SDP

Catalog No. N06455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06455G Supplier TCI America Supplier No. N06455G

missing translation for 'validMainframeMsg'

Specifications

CAS 678-18-2
Color Colorless
Boiling Point 69°C
Molecular Formula C6H3F9O
Quantity 5 g
MDL Number MFCD00153209
UN Number 1224
Synonym 1H,1H,1H-Nonafluoro-2-hexanone, Methyl Perfluorobutyl Ketone, 3,3,4,4,5,5,6,6,6-Nonafluoro-2-hexanone
InChI Key LPPFRPMJUNOKRS-UHFFFAOYSA-N
SMILES CC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexan-2-one
Molecular Weight (g/mol) 262.08
PubChem CID 2775461
Formula Weight 262.07
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonafluorobutyl Ketone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.