Learn More
Medchemexpress LLC Methyl nonanoate | 1731-84-6 | MFCD00009569 | 98.7% | 172.26 g/mol | C10H20O2 | 100 G
SDP
Catalog No. 5000302504
Encompass_Preferred
Methyl nonanoate is the methyl ester of nonanoic (pelargonic) acid, a colorless liquid with a mild fruity odor used as a reagent, reference standard, and fragrance ingredient. It is employed in organic synthesis, analytical chemistry, and formulation work where a medium-chain fatty acid ester is required (CAS 1731-84-6).
  • High purity suitable for analytical use.
  • Typical physical form: colorless liquid with mild fruity odor.
  • Useful as a reference standard for GC and GC-MS analyses.
  • Soluble in common organic solvents for formulation and synthesis.
  • Suitable for esterification and fragrance synthesis applications.
  • Available in multiple laboratory sizes for flexibility.
Catalog No. 50-003-02504 Supplier Medchemexpress LLC Supplier No. HYW015308100G
Methyl nonanoate is the methyl ester of nonanoic (pelargonic) acid, a colorless liquid with a mild fruity odor used as a reagent, reference standard, and fragrance ingredient. It is employed in organic synthesis, analytical chemistry, and formulation work where a medium-chain fatty acid ester is required (CAS 1731-84-6).
  • High purity suitable for analytical use.
  • Typical physical form: colorless liquid with mild fruity odor.
  • Useful as a reference standard for GC and GC-MS analyses.
  • Soluble in common organic solvents for formulation and synthesis.
  • Suitable for esterification and fragrance synthesis applications.
  • Available in multiple laboratory sizes for flexibility.