Learn More
Medchemexpress LLC Methyl nonanoate | 1731-84-6 | MFCD00009569 | 98.7% | 172.27 g/mol | C10H20O2 | 25 G
SDP
Catalog No. 5000303564
Encompass_Preferred
Methyl nonanoate is the methyl ester of nonanoic (pelargonic) acid used as an organic reagent and analytical standard. It is typically a clear liquid with a fruity aroma and is applied in analytical chemistry and flavor and fragrance research as a reference compound.
  • Molecular formula C10H20O2 and molecular weight 172.27 g/mol.
  • High GC purity (98.7% reported on batch certificate of analysis).
  • Suitable as an analytical standard for gas chromatography and other analytical methods.
  • Commonly used in flavor and fragrance research due to its fruity aroma.
  • Offered in laboratory pack sizes suitable for research use.
Catalog No. 50-003-03564 Supplier Medchemexpress LLC Supplier No. 25GHYW015308
Methyl nonanoate is the methyl ester of nonanoic (pelargonic) acid used as an organic reagent and analytical standard. It is typically a clear liquid with a fruity aroma and is applied in analytical chemistry and flavor and fragrance research as a reference compound.
  • Molecular formula C10H20O2 and molecular weight 172.27 g/mol.
  • High GC purity (98.7% reported on batch certificate of analysis).
  • Suitable as an analytical standard for gas chromatography and other analytical methods.
  • Commonly used in flavor and fragrance research due to its fruity aroma.
  • Offered in laboratory pack sizes suitable for research use.