Learn More
Medchemexpress LLC Methyl nonanoate | 1731-84-6 | MFCD00009569 | 98.7% | 172.27 g/mol | C10H20O2 | 500 G
SDP
Catalog No. 5000302626
Encompass_Preferred
Methyl nonanoate is a straight-chain fatty acid methyl ester (CAS 1731-84-6) commonly used as a flavor and fragrance ingredient and as an analytical reagent. It is a colorless to light-yellow liquid with a fruity aroma and is supplied at high purity for laboratory applications.
  • High purity suitable for analytical and research use.
  • Colorless to light-yellow liquid with a fruity odor.
  • Used as a flavoring and fragrance intermediate in formulations.
  • Useful as a solvent or reagent in organic synthesis and analysis.
  • Molecular formula C10H20O2; molecular weight 172.27 g/mol.
Catalog No. 50-003-02626 Supplier Medchemexpress LLC Supplier No. HYW015308500G
Methyl nonanoate is a straight-chain fatty acid methyl ester (CAS 1731-84-6) commonly used as a flavor and fragrance ingredient and as an analytical reagent. It is a colorless to light-yellow liquid with a fruity aroma and is supplied at high purity for laboratory applications.
  • High purity suitable for analytical and research use.
  • Colorless to light-yellow liquid with a fruity odor.
  • Used as a flavoring and fragrance intermediate in formulations.
  • Useful as a solvent or reagent in organic synthesis and analysis.
  • Molecular formula C10H20O2; molecular weight 172.27 g/mol.