Learn More
Medchemexpress LLC Methyl nonanoate (pelargonic acid methyl ester) | 1731-84-6 | MFCD00009569 | 98.7% | 172.27 g/mol | C10H20O2 | 50 G
SDP
Catalog No. 5000302411
Encompass_Preferred
Methyl nonanoate (pelargonic acid methyl ester) is a fatty acid methyl ester with a fruity aroma used as a flavoring and fragrance ingredient, and in industrial applications such as solvents, resins, and plasticizers. It is supplied as a high-purity reagent for laboratory and formulation use.
  • Fruity aroma suitable for flavor and fragrance formulations.
  • High purity for consistent analytical and formulation performance.
  • Wide applicability across food, fragrance, and industrial applications.
  • Available in convenient pack sizes for laboratory and production needs.
  • Liquid ester with defined molecular properties for reproducible results.
Catalog No. 50-003-02411 Supplier Medchemexpress LLC Supplier No. HYW01530850G
Methyl nonanoate (pelargonic acid methyl ester) is a fatty acid methyl ester with a fruity aroma used as a flavoring and fragrance ingredient, and in industrial applications such as solvents, resins, and plasticizers. It is supplied as a high-purity reagent for laboratory and formulation use.
  • Fruity aroma suitable for flavor and fragrance formulations.
  • High purity for consistent analytical and formulation performance.
  • Wide applicability across food, fragrance, and industrial applications.
  • Available in convenient pack sizes for laboratory and production needs.
  • Liquid ester with defined molecular properties for reproducible results.