Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl octanoate

Catalog No. 26067325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-067-325G 25 g
26-067-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-067-325G Supplier Sigma Aldrich Supplier No. 26067325G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl octanoate is a saturated organic compound that belongs to the class of fatty acid methyl ester (fames). it can be prepared by the esterification of octanoic acid with methanol in the presence of an acid catalyst.methyl octanoate can be used as:
  • A reactant to prepare c7 and c8 hydrocarbons by catalytic decarboxylation/decarbonylation reactions in the presence of pt/al2o3 catalyst
  • A component of biodiesel bioethanol surrogate fuel model to study its kinetics of oxidation

Specifications

CAS 111-11-5
Density 0.877 g/mL (at 25°C (literature))
Boiling Point 194°C to 195°C (lit.)
Molecular Formula C9H18O2
Refractive Index n20/D 1.417 (literature)
Linear Formula CH3(CH2)6 COOCH3
MDL Number MFCD00009551
Quantity 25 g
Synonym Caprylic acid methyl ester; Methyl caprylate
Molecular Weight (g/mol) 158.24
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.