Learn More
Medchemexpress LLC Methyl octanoate | 111-11-5 | MFCD00009551 | 99.5% | 158.24 g/mol | C9H18O2 | 1 ML
SDP
Catalog No. 5000208353
Encompass_Preferred
Methyl octanoate is a volatile methyl ester supplied for analytical and research use. It is available as a ready-to-use 10 mM solution in DMSO and in bulk solid quantities for analytical standards and aroma analysis.
  • Ready-to-use 10 mM solution in DMSO
  • Also available in bulk sizes (10 g-500 g)
  • High purity suitable for analytical and flavor analysis
  • CAS number 111-11-5 and molecular weight 158.24 g/mol
  • Used as an aroma component and analytical standard in research
Catalog No. 50-002-08353 Supplier Medchemexpress LLC Supplier No. HYW08794310MM/1ML
Methyl octanoate is a volatile methyl ester supplied for analytical and research use. It is available as a ready-to-use 10 mM solution in DMSO and in bulk solid quantities for analytical standards and aroma analysis.
  • Ready-to-use 10 mM solution in DMSO
  • Also available in bulk sizes (10 g-500 g)
  • High purity suitable for analytical and flavor analysis
  • CAS number 111-11-5 and molecular weight 158.24 g/mol
  • Used as an aroma component and analytical standard in research