Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl octanoate | 111-11-5 | MFCD00009551 | 99.6% | 158.24 g/mol | C9H18O2 | 25 G
SDP

Catalog No. 5000209621
Encompass_Preferred

Methyl octanoate is a volatile fatty acid methyl ester used as an aroma component and as an analytical standard for research and laboratory applications. It is supplied as a high-purity liquid in multiple pack sizes for analytical or preparative use.

  • volatile fatty acid methyl ester with fruity/ester aroma
  • used as an analytical standard and research reagent
  • high purity (approximately 99.5% by chromatography)
  • molecular weight 158.24 g/mol and formula C9H18O2
  • available in small solution formats and solid pack sizes up to bulk quantities
  • physical form: colorless liquid (ester)

Catalog No. 50-002-09621 Supplier Medchemexpress LLC Supplier No. 25GHYW087943
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl octanoate is a volatile fatty acid methyl ester used as an aroma component and as an analytical standard for research and laboratory applications. It is supplied as a high-purity liquid in multiple pack sizes for analytical or preparative use.

  • volatile fatty acid methyl ester with fruity/ester aroma
  • used as an analytical standard and research reagent
  • high purity (approximately 99.5% by chromatography)
  • molecular weight 158.24 g/mol and formula C9H18O2
  • available in small solution formats and solid pack sizes up to bulk quantities
  • physical form: colorless liquid (ester)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.