Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl octanoate | 111-11-5 | MFCD00009551 | 99.6% | 158.24 g·mol⁻¹ | C9H18O2 | 50 G
SDP

Catalog No. 5000208332
Encompass_Preferred

Methyl octanoate is the methyl ester of octanoic (caprylic) acid, a volatile organic ester used as an aroma component and analytical reference. Supplied as a high-purity liquid for laboratory and analytical use; not for human consumption.

  • CAS number: 111-11-5.
  • Molecular formula: C9H18O2.
  • Molecular weight: 158.24 g·mol⁻¹.
  • Purity: 99.6%.
  • Form: liquid.
  • Intended use: research and analytical applications; not for human consumption.
  • Typical package sizes: 10 g, 25 g, 50 g, 100 g, 500 g.

Catalog No. 50-002-08332 Supplier Medchemexpress LLC Supplier No. HYW08794350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl octanoate is the methyl ester of octanoic (caprylic) acid, a volatile organic ester used as an aroma component and analytical reference. Supplied as a high-purity liquid for laboratory and analytical use; not for human consumption.

  • CAS number: 111-11-5.
  • Molecular formula: C9H18O2.
  • Molecular weight: 158.24 g·mol⁻¹.
  • Purity: 99.6%.
  • Form: liquid.
  • Intended use: research and analytical applications; not for human consumption.
  • Typical package sizes: 10 g, 25 g, 50 g, 100 g, 500 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.