Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl octanoate, EMD-Millipore

Catalog No. M1096330005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096330005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096330005 Supplier MilliporeSigma Supplier No. 1.09633.0005

missing translation for 'validMainframeMsg'

Fatty acid methyl esters

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-11-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
InChI Key JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonym Methyl caprylate
PubChem CID 8091
ChEBI CHEBI:87432
IUPAC Name methyl octanoate
SMILES CCCCCCCC(=O)OC

Specifications

Packaging Glass vials
Vapor Pressure 1.33 hPa (34.2°C)
Density Density: 0.877g/cm3 (20°)
Assay Percent Range ≥99.5%
Quantity 5 mL
Formula Weight 158.24
Chemical Name or Material Methyl octanoate
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.