Learn More
Medchemexpress LLC Methyl oleate | 112-62-9 | 1 G
SDP
Catalog No. 5000420260
Encompass_Preferred
Methyl oleate is a fatty acid methyl ester (FAME) that exhibits anti-extended spectrum β-Lactamase (ESBL) potential, as well as anticancer and antibacterial activities. It enhances the antioxidation ability but impairs the antiwear capacity of zinc dialkyldithiophosphate (ZDDP). This compound is considered promising for research involving ESBL-producing multi-drug resistant (MDR) pathogens.
  • Exhibits anti-extended spectrum β-lactamase (ESBL) potential
  • Exhibits anticancer and antibacterial activities
  • Enhances the antioxidation ability of zinc dialkyldithiophosphate (ZDDP)
  • Promising for research involving ESBL-producing multi-drug resistant (MDR) pathogens
  • Inhibits ESBL producing MDR *E. coli* and *K. pneumoniae*
  • Inhibits A549 cells with an IC50 value of 625 μg/mL
Catalog No. 50-004-20260 Supplier Medchemexpress LLC Supplier No. HYN25981G
Methyl oleate is a fatty acid methyl ester (FAME) that exhibits anti-extended spectrum β-Lactamase (ESBL) potential, as well as anticancer and antibacterial activities. It enhances the antioxidation ability but impairs the antiwear capacity of zinc dialkyldithiophosphate (ZDDP). This compound is considered promising for research involving ESBL-producing multi-drug resistant (MDR) pathogens.
  • Exhibits anti-extended spectrum β-lactamase (ESBL) potential
  • Exhibits anticancer and antibacterial activities
  • Enhances the antioxidation ability of zinc dialkyldithiophosphate (ZDDP)
  • Promising for research involving ESBL-producing multi-drug resistant (MDR) pathogens
  • Inhibits ESBL producing MDR *E. coli* and *K. pneumoniae*
  • Inhibits A549 cells with an IC50 value of 625 μg/mL