Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl oleate | 112-62-9 | 1 ML
SDP

Catalog No. 5000420463
Encompass_Preferred

Methyl oleate is a fatty acid methyl ester (FAME) that exhibits several biological activities, including anti-extended spectrum β-Lactamase (ESBL) potential, anticancer, and antibacterial properties. It has been observed to improve the antioxidation ability of zinc dialkyldithiophosphate (ZDDP) while simultaneously impairing its antiwear capacity. This compound holds promise for research involving ESBL-producing multi-drug resistant (MDR) pathogens.

  • Anticancer activity: Inhibits A549 cells (IC50 of 625 μg/mL after 48 hours), affecting cell viability by inhibiting cell-to-cell contact.
  • Antibacterial activity: Effective against ESBL-producing MDR E. coli and K. pneumoniae (80 and 100 μg/mL).
  • Research potential: Promising for studies on ESBL-producing multi-drug resistant (MDR) pathogens.
  • Physical characteristics: Colorless to off-white liquid with a density of 0.874 g/cm³.
  • Purity: 99.57%.

Catalog No. 50-004-20463 Supplier Medchemexpress LLC Supplier No. HYN259810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl oleate is a fatty acid methyl ester (FAME) that exhibits several biological activities, including anti-extended spectrum β-Lactamase (ESBL) potential, anticancer, and antibacterial properties. It has been observed to improve the antioxidation ability of zinc dialkyldithiophosphate (ZDDP) while simultaneously impairing its antiwear capacity. This compound holds promise for research involving ESBL-producing multi-drug resistant (MDR) pathogens.

  • Anticancer activity: Inhibits A549 cells (IC50 of 625 μg/mL after 48 hours), affecting cell viability by inhibiting cell-to-cell contact.
  • Antibacterial activity: Effective against ESBL-producing MDR E. coli and K. pneumoniae (80 and 100 μg/mL).
  • Research potential: Promising for studies on ESBL-producing multi-drug resistant (MDR) pathogens.
  • Physical characteristics: Colorless to off-white liquid with a density of 0.874 g/cm³.
  • Purity: 99.57%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.