Learn More
Medchemexpress LLC Methyl oleate | 112-62-9 | 500 MG
SDP
Catalog No. 5000420772
Encompass_Preferred
Methyl oleate is a fatty acid methyl ester (FAME) that exhibits anti-extended spectrum β-Lactamase (ESBL) potential, as well as anticancer and antibacterial activities. It significantly improves antioxidation ability but impairs the antiwear capacity of zinc dialkyldithiophosphate (ZDDP). This substance shows promise for research involving ESBL-producing multi-drug resistant (MDR) pathogens.
  • Effective in inhibiting ESBL-producing MDR E. coli and K. pneumoniae in vitro
  • Exerts inhibition on A549 cells in vitro with an IC50 value of 625 μg/mL
  • Demonstrated efficacy against specific bacterial strains
  • Substantially enhances antioxidation ability
  • Promising for studies on ESBL-producing multi-drug resistant (MDR) pathogens
  • Induced behavioral responses in Lobeo rohita, such as hyperventilation and loss of reflex, at lethal concentrations
Catalog No. 50-004-20772 Supplier Medchemexpress LLC Supplier No. HYN2598500MG
Methyl oleate is a fatty acid methyl ester (FAME) that exhibits anti-extended spectrum β-Lactamase (ESBL) potential, as well as anticancer and antibacterial activities. It significantly improves antioxidation ability but impairs the antiwear capacity of zinc dialkyldithiophosphate (ZDDP). This substance shows promise for research involving ESBL-producing multi-drug resistant (MDR) pathogens.
  • Effective in inhibiting ESBL-producing MDR E. coli and K. pneumoniae in vitro
  • Exerts inhibition on A549 cells in vitro with an IC50 value of 625 μg/mL
  • Demonstrated efficacy against specific bacterial strains
  • Substantially enhances antioxidation ability
  • Promising for studies on ESBL-producing multi-drug resistant (MDR) pathogens
  • Induced behavioral responses in Lobeo rohita, such as hyperventilation and loss of reflex, at lethal concentrations