Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl oleate

Catalog No. 3111115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-111-15G 5 g
31-111-1250G 250 g
31-111-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-111-15G Supplier Sigma Aldrich Supplier No. 3111115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The methyl oleate monolayers at the air-water interface undergoes ozonolysis that results in rapid loss of material through cleavage of the C=C bond and evaporation/dissolution of reaction products.Methyl oleate was used to study the catalytic potential of molybdenum oxide/bipyridinedicarboxylate hybrid material.

Specifications

CAS 112-62-9
Density 0.874 g/mL (at 20°C (literature))
Boiling Point 218°C (20 mmHg)
Molecular Formula C19H36O2
Linear Formula CH3(CH2)7 CH=CH(CH2)7 CO2CH3
Refractive Index n20/D 1.452 (literature)
MDL Number MFCD00009578
Quantity 5 g
Synonym Methyl cis-9-octadecenoate; Oleic acid methyl ester
Molecular Weight (g/mol) 296.49
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.