Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl oleate 99% | 112-62-9 | MFCD00009578 | 5G
Catalog No. NC2147406
Encompass
Methyl oleate 99% | Purity: 99% | Mol Wt: 296.49 | 112-62-9 | MFCD00009578 | 5G
Catalog No. NC2147406 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3111115G
Methyl oleate 99% | Purity: 99% | Mol Wt: 296.49 | 112-62-9 | MFCD00009578 | 5G