Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl oleate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012501 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2501 5 mL
11-101-2500 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2501 Supplier MilliporeSigma Supelco Supplier No. 751605ML
Add to Cart
Add to Cart

Methyl oleate can be obtained as one of the products of transesterification of triolein with methanol in the presence of potassium carbonate loaded onto alumina as catalyst.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 112-62-9
Vapor Pressure 10 mmHg (205°C)
Density 0.874 g/mL (at 20°C)
Flash Point 113.0°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)7 CO2 CH3
MDL Number MFCD00009578
Boiling Point 218°C (20 mmHg)
Synonym Methyl cis-9-octadecenoate; Oleic acid methyl ester
Type Analytical Standard
Quantity 5 mL
Formula Weight 296.49
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.