Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl oleate, technical

Catalog No. ac414645000
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414645000 500 g Glass bottle
AC414640010 1 kg Glass bottle
AC414640050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414645000 Supplier Thermo Scientific Chemicals Supplier No. 414645000
Only null left
Add to Cart

CAS: 67762-38-3 | C19H36O2 | 296.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67762-38-3
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.50
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-ZHACJKMWSA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -20.0°C
Color Yellow to Amber
Density 0.8700g/mL
Boiling Point 186.0°C (9.0 mmHg)
Flash Point 180°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4510 to 1.4580
Linear Formula CH3(CH2)7CHCH(CH2)7COOCH3
Quantity 500 g
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Viscosity 6.829 mPa.s (20°C)
Formula Weight 296.48
Percent Purity ≥55%
Grade Technical
Physical Form Oily Liquid
Chemical Name or Material Methyl oleate
Show More Show Less
EINECSNumber 203-992-5
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.