Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange, 0.1% (w/v) Aqueous Indicator Solution, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog No. 18604433 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-604-433 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-433 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M218500MLPL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M-218, 547-58-0

Spectrum™ Chemical Methyl Orange, 0.1 Percent (w/v) Aqueous Indicator Solution, (U.S.P. Test Solution) is a Spectrum solution concentrate that allows the preparation of fresh, reliable standard volumetric solutions quickly and accurately. Simply dilute the desired USP test solution concentrate to the specified volume (carefully following instructions provided with the package) and the resulting solution will have an exact concentration within 0.1%. Spectrum USP test solution concentrates are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications.

Chemical Identifiers

CAS 547-58-0 , 7732-18-5
Molecular Formula C14H14N3O3S
Molecular Weight (g/mol) 304.34
InChI Key IETWCRRCPURZOC-UHFFFAOYSA-M
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Max % 0.1%, 99.9%
Concentration 0.1%
Packaging Poly Bottle
Color Orange
Quantity 500 mL
Physical Form Chemical Solution
Grade Chemical Solution
Chemical Name or Material Methyl Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.