Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange, 0.2% Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18611712 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-611-712 100 mL
18-611-713 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-712 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M340100ML
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Methyl Orange
CAS 547-58-0, 7732-18-5
Concentration 0.2 Percent
Grade Chemical Solution
Packaging Poly Bottle
Color Orange
Quantity 100 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.