Learn More
Hach Company Methyl Orange Indicator Solution, 100 mL MDB
Supplier:  Hach Company 14832
Encompass
pH Indicator for acidity titrations. 0.50 g/L, APHA formulation. Color change pink (pH 3.2) to yellow (pH 4.4). 100 mL Marked Dropping Bottle
Catalog No. 50-199-4177