Learn More
Medchemexpress LLC Methyl orange | 547-58-0 | MFCD00007502 | 99.9% | 327.33 g/mol | C14H14N3NaO3S | 50 G
SDP
Catalog No. 5000228585
Encompass_Preferred
Methyl orange is a water-soluble azo dye supplied as a solid powder and used primarily as an acid-base indicator and a staining or dyeing reagent. It exhibits a color change from red to yellow across the pH 3.1-4.4 range, making it useful for titration end-point detection and analytical staining.
  • High purity reported at 99.85%.
  • Distinct color transition at pH 3.1-4.4 for accurate end-point detection.
  • Solid powder form suitable for preparing indicator solutions or dyes.
  • Common applications include titration, tissue staining, and textile dyeing.
  • Handle with care; may be cytotoxic at high concentrations.
Catalog No. 50-002-28585 Supplier Medchemexpress LLC Supplier No. HY11890750G
Add to Cart
Add to Cart
Methyl orange is a water-soluble azo dye supplied as a solid powder and used primarily as an acid-base indicator and a staining or dyeing reagent. It exhibits a color change from red to yellow across the pH 3.1-4.4 range, making it useful for titration end-point detection and analytical staining.
  • High purity reported at 99.85%.
  • Distinct color transition at pH 3.1-4.4 for accurate end-point detection.
  • Solid powder form suitable for preparing indicator solutions or dyes.
  • Common applications include titration, tissue staining, and textile dyeing.
  • Handle with care; may be cytotoxic at high concentrations.